Skip to Content
Merck
All Photos(1)

Key Documents

334618

Sigma-Aldrich

trans-Diamminetetrachloroplatinum(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)2Cl4
CAS Number:
Molecular Weight:
370.95
MDL number:
UNSPSC Code:
12352302

mp

>300 °C (lit.)

SMILES string

N.N.Cl[Pt](Cl)(Cl)Cl

InChI

1S/4ClH.2H3N.Pt/h4*1H;2*1H3;/q;;;;;;+4/p-4

InChI key

RBHOYUZKIANPPG-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Plooy et al.
Cancer research, 44(5), 2043-2051 (1984-05-01)
Several effects of four diamminechloroplatinum compounds (II and IV) in Chinese hamster ovary cells were studied. The two cis-compounds [cis-diamminedichloroplatinum(II) and cis-diamminetetrachloroplatinum(IV)] are known to possess antitumor activity, whereas the two trans-stereoisomers [trans-diamminedichloroplatinum(II) and trans-diamminetetrachloroplatinum(IV)] are inactive. When the effects
M Defais et al.
Chemico-biological interactions, 74(3), 343-352 (1990-01-01)
DNA strand breaks were observed in bacteria treated with Pt(IV) but not Pt(II) antitumor compounds by two methods. First, compounds which cause DNA strand breaks produced an SOS induction signal which was detected by a rapid bacterial assay. In addition
H Razaka et al.
Chemico-biological interactions, 60(2), 207-215 (1986-11-01)
After exposure of bacteria to equal concentrations of cis-diamminedichloroplatinum(II) (DDP) and cis-diamminetetrachloroplatinum(IV) (DTP), the intracellular concentration of DTP was an order of magnitude greater than DDP. However, at identical intracellular drug concentrations, the Pt(IV) compound formed only half as many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service