Skip to Content
Merck
All Photos(3)

Documents

333948

Sigma-Aldrich

Zirconium(IV) butoxide solution

80 wt. % in 1-butanol

Synonym(s):

Tetrabutyl zirconate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(OC4H9)4
CAS Number:
Molecular Weight:
383.68
Beilstein:
4132649
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solution

reaction suitability

core: zirconium
reagent type: catalyst

concentration

80 wt. % in 1-butanol

refractive index

n20/D 1.466

density

1.049 g/mL at 25 °C

SMILES string

CCCCO[Zr](OCCCC)(OCCCC)OCCCC

InChI

1S/4C4H9O.Zr/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4

InChI key

BSDOQSMQCZQLDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in an unconventional method to prepare a gel of CeO2-stabilized zirconia polycrystals that have promising applications as protective coatings of steel and ferrous alloys.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DiMaggio, R. et al.
Chemistry of Materials, 10, 3839-3839 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service