Skip to Content
Merck
All Photos(1)

Documents

333751

Sigma-Aldrich

Potassium cyanide-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KC15N
CAS Number:
Molecular Weight:
66.11
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

form

solid

mp

>300 °C (lit.)

mass shift

M+1

SMILES string

[K]C#[15N]

InChI

1S/CN.K/c1-2;/i2+1;

InChI key

YUZRZFQHUCKACF-CGOMOMTCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Zhang Yu et al.
Journal of hazardous materials, 225-226, 190-194 (2012-05-29)
A study was conducted to investigate activities of nitrate reductase (NR) and glutamine synthetase (GS) in plants during cyanide metabolism. Young rice seedlings (Oryza sativa L. cv. XZX 45) were grown in the nutrient solutions containing KNO(3) or NH(4)Cl and
Anand Kumar et al.
Amino acids, 43(6), 2417-2429 (2012-05-23)
Condensation reactions of amino acid (glycine and alanine) on the surface of metal(II) octacyanomolybdate(IV) (MOCMo) complexes are investigated using high-performance liquid chromatography (HPLC) and electron spray ionizations-mass spectroscopy (ESI-MS). The series of MOCMo have been synthesized and the effect of
Erica M Granjeiro et al.
Autonomic neuroscience : basic & clinical, 167(1-2), 21-26 (2011-12-27)
Several studies from our group have indicated that the BNST play an important role in baroreflex modulation. However, the involvement of the BNST in the chemoreflex activity is unknown. Thus, in the present study, we investigated the effect of the
Naresh K Sharma et al.
Bioresource technology, 121, 263-273 (2012-08-04)
Present study focused on the degradation of a mixture of phenol, cresol, xylenol, quinoline, and indole along with cyanide, commonly found in coke oven wastewater, using aerobic mixed culture. It was found that xylenol and indole were difficult to degrade
Fei Xu et al.
Plant, cell & environment, 35(11), 1983-1997 (2012-05-05)
The effects of potassium cyanide (KCN) pretreatment on the response of cucumber (Cucumis sativus L.) plants to salt, polyethylene glycol (PEG) and cold stress were investigated in the present study. Here, we found that KCN pretreatment improved cucumber seedlings tolerance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service