Skip to Content
Merck
All Photos(1)

Key Documents

333433

Sigma-Aldrich

1,2-Bis(2-iodoethoxy)ethane

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€56.40

€56.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€56.40

About This Item

Linear Formula:
ICH2CH2OCH2CH2OCH2CH2I
CAS Number:
Molecular Weight:
369.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€56.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.572 (lit.)

density

2.028 g/mL at 25 °C (lit.)

functional group

ether
iodo

SMILES string

ICCOCCOCCI

InChI

1S/C6H12I2O2/c7-1-3-9-5-6-10-4-2-8/h1-6H2

InChI key

BCAGFJXMCZSAHD-UHFFFAOYSA-N

Related Categories

Application

1,2- Bis (2-iodoethoxy)ethane was used as crosslinking reagent for synthesis of:
  • zirconia-based, alkali-stable strong anion-exchange stationary phase[1]
  • silica-coated nanoparticles[2]
  • novel shell cross-linked (SCL) micelles with pH-responsive poly(2-(diethylamino)ethyl methacrylate) cores and thermoresponsive poly(N-isopropylacrylamide) coronas[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuting Li et al.
Macromolecular rapid communications, 30(6), 464-468 (2009-03-19)
The mean diameter of poly[2-(dimethylamino)ethyl methacrylate]-block-poly[2-(diisopropylamino)ethyl methacrylate] (PDMA-PDPA) diblock copolymer micelles can be easily adjusted from 27-155 nm (as measured by DLS) by either selective quaternisation of the PDMA block or by adding PDPA homopolymer prior to micellisation; these self-assembled
C McNeff et al.
Analytical biochemistry, 274(2), 181-187 (1999-10-21)
The synthesis and use of a zirconia-based, alkali-stable strong anion-exchange stationary phase are described for the removal of pyrogenic lipopolysaccharides (LPS) from insulin. The strong anion-exchange material is produced by deposition of polyethyleneimine (PEI) onto porous zirconia particles, followed by
Xiaoze Jiang et al.
Biomacromolecules, 8(10), 3184-3192 (2007-09-25)
A double hydrophilic ABC triblock copolymer, poly(2-(diethylamino)ethyl methacrylate)-b-poly(2-(dimethylamino)ethyl methacrylate)-b-poly(N-isopropylacrylamide) (PDEA-b-PDMA-b-PNIPAM), containing the well-known pH-responsive PDEA block and thermoresponsive PNIPAM block, was synthesized by atom transfer radical polymerization via sequential monomer addition using ethyl 2-chloropropionate as the initiator. The obtained triblock
Yueyuan Mao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(46), 16623-16630 (2015-09-30)
Two large conjugated naphthalimide derivatives with or without three-methane-bridged thiazole orange (TO3; i.e., compounds 1 a and 2 a, respectively) were designed and synthesized. The fluorescence of the naphthalimide group in compound 1 a at λ=532 nm initially decreased and that for the TO3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service