Skip to Content
Merck
All Photos(2)

Key Documents

333212

Sigma-Aldrich

Thallium(I) carbonate

99.9% trace metals basis

Synonym(s):

Thallous carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€76.10

€76.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€76.10

About This Item

Linear Formula:
Tl2CO3
CAS Number:
Molecular Weight:
468.78
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystalline

€76.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.9% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: thallium

impurities

≤1500.0 ppm Trace Metal Analysis

mp

260-270 °C (dec.) (lit.)

density

7.11 g/mL at 25 °C (lit.)

SMILES string

[Tl]OC(=O)O[Tl]

InChI

1S/CH2O3.2Tl/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

DASUJKKKKGHFBF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Studies on the spermotoxicity of thallium carbonate in drinking water and its effect on the reproductive function of mice].
Q Y Wei
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 21(3), 141-143 (1987-05-01)
[Experimental substantiation of maximum permissible concentration of thallium carbonate in environmental air].
A Kh Kamil'dzhanov
Gigiena i sanitariia, (5)(5), 8-10 (1993-05-01)
T S Thian et al.
Applied and environmental microbiology, 41(3), 724-728 (1981-03-01)
Gentamicin-thallous-carbonate (GTC) agar was formulated by Donnelly and Hartman (Appl. Environ. Microbiol. 35:576-581, 1978) to select for fecal streptococci in sewage and water samples. The present study was conducted to determine the usefulness of GTC agar for the enumeration of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service