Skip to Content
Merck
All Photos(1)

Key Documents

332844

Sigma-Aldrich

4-Methoxycyclohexanecarboxylic acid, mixture of cis and trans

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H10CO2H
CAS Number:
Molecular Weight:
158.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4687 (lit.)

functional group

carboxylic acid
ether

SMILES string

COC1CCC(CC1)C(O)=O

InChI

1S/C8H14O3/c1-11-7-4-2-6(3-5-7)8(9)10/h6-7H,2-5H2,1H3,(H,9,10)

InChI key

WKILSRYNRQGRMA-UHFFFAOYSA-N

General description

cis-4-Methoxycyclohexanecarboxylic acid reacts with methyllithium to afford ketone, cis-4-methoxy-1-acetylcyclohexane.[1] Reaction of cis-4-methoxycyclohexanecarboxylic acid with acetic acid and catalytic amount of sulphuric acid has been investigated.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes-Studies of Configuration. VI. cis-and trans-4-Methoxycyclohexanol.
Noyce DS and Weingarten HI.
The Journal of Organic Chemistry, 25(2), 260-262 (1960)
Studies of Configuration. IV. The Rearrangement of Methoxycyclohexanecarboxylic Acids with Acetic Anhydride1.
Noyce DS and Weingarten HI.
Journal of the American Chemical Society, 79(12), 3098-3102 (1957)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service