Skip to Content
Merck
All Photos(2)

Key Documents

330507

Sigma-Aldrich

Ethylenediaminetetraacetic acid diammonium salt hydrate

97%

Synonym(s):

Diammonium EDTA monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€32.70
100 G
€87.80

€32.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€32.70
100 G
€87.80

About This Item

Linear Formula:
HO2CCH2N(CH2CO2NH4)CH2CH2N(CH2CO2NH4)CH2CO2H·xH2O
CAS Number:
Molecular Weight:
326.30 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€32.70


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

218 °C (dec.) (lit.)

functional group

amine
carboxylic acid

SMILES string

N.N.O.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.2H3N.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);2*1H3;1H2

InChI key

WRSUMHYBOYWMTD-UHFFFAOYSA-N

General description

Ethylenediaminetetraacetic acid diammonium salt hydrate, also known as EDTA diammonium salt hydrate. It is a derivative of ethylenediaminetetraacetic acid (EDTA) and is commonly used as a chelating agent in various applications due to its ability to bind to various metal ions and form stable complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsushi Hirano et al.
ACS nano, 13(2), 1805-1816 (2019-01-27)
Nanomaterials introduced into biological systems are immediately coated by proteins in vivo. They induce oxidative stress on adsorbed proteins and hence alter the protein structures, which determines the fate pathways and biological impacts of nanomaterials. Carbon nanotubes (CNTs) have been

Questions

  1. The Cas number 304675-80-7 for diammonium EDTA indicates only one water of hydration, but when we prepared a solution, the titrated strength was much less suggesting a higher number of waters of hydration. What is the maximum value of X in xH2O?

    1 answer
    1. The CAS number is associated with the 'hydrate' form of this compound. The water content may be variable. The maximum water content allowable for this product is 5%. The water content is lot specific and reported in the product Certificate of Analysis. Please see the link below to access a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/aldrich/330507#product-documentation
      Please see the link below to review the PubChem listing for this CAS number:
      https://pubchem.ncbi.nlm.nih.gov/compound/71311375

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service