Skip to Content
Merck
All Photos(2)

Key Documents

330094

Sigma-Aldrich

Palladium hydroxide on carbon

extent of labeling: 20 wt. % loading (dry basis), matrix carbon, wet support

Synonym(s):

Pearlman’s catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€263.00
50 G
€1,250.00

€263.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€263.00
50 G
€1,250.00

About This Item

Empirical Formula (Hill Notation):
H2O2Pd
CAS Number:
Molecular Weight:
140.43
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€263.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

contains

~50% water

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

20 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

carbon, wet support

SMILES string

O[Pd]O

InChI

1S/2H2O.Pd/h2*1H2;/q;;+2/p-2

InChI key

NXJCBFBQEVOTOW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogenolysis of benzyl groups and epoxides; used to reduce nitro compounds to amines and as dehydrogenation reagent.
Useful reagent for debenzylation.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Nie et al.
European journal of mass spectrometry (Chichester, England), 21(3), 287-296 (2015-08-27)
Complex mixtures of plant derived triglycerol (TG) lipids are commonly used as feedstock components for the production of industrial polymers. However, there remains a need for the development of analytical strategies to investigate the intrinsic intermolecular cross-linking reactivity of individual
Yossi Shemesh et al.
Nucleosides, nucleotides & nucleic acids, 34(11), 753-762 (2015-10-01)
Carboxylates and dicarboxylates are important phosphate mimics. Herein, we present a simple synthetic route for the preparation of RNA carboxylate/dicarboxylate conjugates, starting from suitably protected NH2- and COOH-containing molecules that are coupled to the RNA on the solid support. The

Articles

An application guide to help you select the most suitable heterogeneous catalysts for diverse synthetic applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service