Skip to Content
Merck
All Photos(1)

Documents

328731

Sigma-Aldrich

2-Nitrophenyl chloroformate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2C6H4NO2
CAS Number:
Molecular Weight:
201.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

bp

132-133 °C/7 mmHg (lit.)

mp

35-38 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1OC(Cl)=O

InChI

1S/C7H4ClNO4/c8-7(10)13-6-4-2-1-3-5(6)9(11)12/h1-4H

InChI key

FHLXUWOHGKLDNF-UHFFFAOYSA-N

Other Notes

remainder 2-nitrophenol

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofei Wang et al.
International journal of nanomedicine, 13, 4641-4659 (2018-08-28)
Improvement in drug accumulation in the lungs through inhalation administration and high expression of MMP2 and MMP9 in lung tumors have both been widely reported. MMP2/9-triggered-release micelles were constructed and in vitro and in vivo studies of inhalation administration against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service