Skip to Content
Merck
All Photos(2)

Key Documents

325783

Sigma-Aldrich

Lutetium(III) acetate hydrate

99.9% trace metals basis

Synonym(s):

Lutetium triacetate, Lutetium(3+) triacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€103.00

About This Item

Linear Formula:
(CH3CO2)3Lu · xH2O
CAS Number:
Molecular Weight:
352.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€103.00


Check Cart for Availability

Request a Bulk Order

Assay

99.9% trace metals basis

form

powder or crystals

reaction suitability

core: lutetium
reagent type: catalyst

SMILES string

O.CC(=O)O[Lu](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Lu.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

ZMTOUBHBKDQYAB-UHFFFAOYSA-K

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianying Sun et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(5), 766-770 (2015-10-01)
Surface coating is a commonly used strategy to enhance upconversion emissions by shielding the luminescent core from surface quenching. In this work, we provide insights into the effect of surface coating on upconversion by investigating NaYF4 :Yb/Er nanoparticles and the
Kyungseop Lim et al.
Pharmaceutics, 12(11) (2020-11-21)
Photodynamic therapy (PDT) combined with upconverting nanoparticles (UCNPs) are viewed together as an effective method of ablating tumors. After absorbing highly tissue-penetrating near-infrared (NIR) light, UCNPs emit a shorter wavelength light (~660 nm) suitable for PDT. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service