Skip to Content
Merck
All Photos(2)

Key Documents

325058

Sigma-Aldrich

(S)-(+)-3-Bromo-2-methyl-1-propanol

97%

Synonym(s):

(2S)-3-Bromo-2-methyl-1-propanol, (S)-3-Bromo-2-methylpropan-1-ol, (S)-3-Hydroxy-2-methylpropyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
153.02
Beilstein:
4738280
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]25/D +7.3°, c = 2 in chloroform

refractive index

n20/D 1.483 (lit.)

density

1.461 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

C[C@@H](CO)CBr

InChI

1S/C4H9BrO/c1-4(2-5)3-6/h4,6H,2-3H2,1H3/t4-/m1/s1

InChI key

KIBOHRIGZMLNNS-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-3-Bromo-2-methyl-1-propanol is a precursor to synthesize the phase II mexiletine metabolite, (R)-mexiletine N-carbonyloxy-β-D-glucuronide.[1] It can be used in the synthesis of (R)-(+)-muscopyridine,[2] polycavernoside A[3] and (+)-allopumiliotoxin 323B′.[4] It can also be employed in building homochiral porous molecular networks.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of the marine toxin polycavernoside A via selective macrolactonization of a trihydroxy carboxylic acid.
White J d, et al.
Journal of the American Chemical Society, 123(35), 8593-8595 (2001)
A Catalytic Approach to (R)?(+)?Muscopyridine with Integrated ?Self?Clearance?.
Furstner A and Leitner A
Angewandte Chemie (International Edition in English), 42(3), 308-311 (2003)
First synthesis and full characterization of mexiletine N-carbonyloxy ?-D-glucuronide.
Cavalluzzi M M, et al.
Tetrahedron Letters, 51(40), 5265-5268 (2010)
The Synthesis of (+)?Allopumiliotoxin 323B?.
Tan CH and Holmes A B
Chemistry?A European Journal , 7(9), 1845-1854 (2001)
Control and induction of surface-confined homochiral porous molecular networks.
Tahara K, et al.
Nature Chemistry, 3(9), 714-719 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service