All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
liquid
refractive index
n20/D 1.5479 (lit.)
bp
88 °C/1.3 mmHg (lit.)
density
1.055 g/mL at 25 °C (lit.)
functional group
nitrile
SMILES string
N#CC1Cc2ccccc12
InChI
1S/C9H7N/c10-6-8-5-7-3-1-2-4-9(7)8/h1-4,8H,5H2
InChI key
FJIDKRPZJBUHME-UHFFFAOYSA-N
General description
Enantiomeric resolution of 1-benzocyclobutenecarbonitrile by CHIRALPAK IA, CHIRALPAK IB and CHIRALPAK IC (new generation chiral stationary phases) by chromatography was reported.
Application
1-Benzocyclobutenecarbonitrile was used as precursors of R-cyano- or R-keto-substituted o-quinodimethanes.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tong Zhang et al.
Journal of chromatography. A, 1191(1-2), 214-222 (2008-01-08)
CHIRALPAK IA, CHIRALPAK IB and CHIRALPAK IC are a new generation of chiral packing materials for resolution of enantiomers by chromatography. They are all prepared by chemical immobilisation of the respective polysaccharide derivatives onto a supporting silica matrix. The present
o-Quinodimethanes: Efficient Intermediates in Organic Synthesis.
J L Segura et al.
Chemical reviews, 99(11), 3199-3246 (2001-12-26)
Diane L Haakonsen et al.
Nature, 626(8000), 874-880 (2024-02-01)
Stress response pathways detect and alleviate adverse conditions to safeguard cell and tissue homeostasis, yet their prolonged activation induces apoptosis and disrupts organismal health1-3. How stress responses are turned off at the right time and place remains poorly understood. Here
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service