Skip to Content
Merck
All Photos(1)

Key Documents

323233

Sigma-Aldrich

Oxalyl bromide solution

2.0 M in methylene chloride

Synonym(s):

Ethanedioyl dibromide solution, NSC 96957

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCOCOBr
CAS Number:
Molecular Weight:
215.83
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

reagent type: oxidant

concentration

2.0 M in methylene chloride

density

1.517 g/mL at 25 °C

functional group

acyl bromide

SMILES string

BrC(=O)C(Br)=O

InChI

1S/C2Br2O2/c3-1(5)2(4)6

InChI key

JAZLVNXWYDFQFE-UHFFFAOYSA-N

Application

Used in synthetic applications of carbon-substituted iminium salts

Reactant for:
  • Synthesis of mutasynthons added to cultures of A. pretiosum for mutasynthetic generation of ansamitocin derivatives
  • Oxalic acid formation from hydroxyl radical substitutions
  • Cyclization to produce CRF1 receptor antagonists
  • Preparation, optical and electrochemical studies of thiophene end capped olig(2,3-alkylthieno[3,4-b]pyrazine)
  • Asymmetrical synthesis of glycosyl chlorides and bromides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service