Skip to Content
Merck
All Photos(2)

Key Documents

320587

Sigma-Aldrich

Benzyl cyanoformate

97%

Synonym(s):

Cyanoformic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCO2CH2C6H5
CAS Number:
Molecular Weight:
161.16
Beilstein:
1947475
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.505 (lit.)

bp

66-67 °C/0.6 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

SMILES string

O=C(OCc1ccccc1)C#N

InChI

1S/C9H7NO2/c10-6-9(11)12-7-8-4-2-1-3-5-8/h1-5H,7H2

InChI key

GDZIODIYBLTRRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service