Skip to Content
Merck
All Photos(1)

Key Documents

320374

Sigma-Aldrich

Methacryloyl chloride

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COCl
CAS Number:
Molecular Weight:
104.53
Beilstein:
878175
MDL number:
Pricing and availability is not currently available.

grade

technical grade

Assay

90%

contains

200 ppm phenothiazine as stabilizer

refractive index

n20/D 1.442 (lit.)

bp

95-96 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(Cl)=O

InChI

1S/C4H5ClO/c1-3(2)4(5)6/h1H2,2H3

InChI key

VHRYZQNGTZXDNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in poly-coated bottle

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service