Skip to Content
Merck
All Photos(1)

Key Documents

318388

Sigma-Aldrich

Hexadecanophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO(CH2)14CH3
CAS Number:
Molecular Weight:
316.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

bp

248 °C/12 mmHg (lit.)

mp

61-65 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC(=O)c1ccccc1

InChI

1S/C22H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-17-20-22(23)21-18-15-14-16-19-21/h14-16,18-19H,2-13,17,20H2,1H3

InChI key

IIOLAWJMOGLOIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James P Grinias et al.
Journal of chromatography. A, 1371, 261-264 (2014-12-03)
At high flow rates and pressures, columns packed with sub-2 μm particles suffer from efficiency losses due to frictional heating. The thermal environment of the column (insulated or isothermal) can decrease or magnify these losses. While a number of studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service