Skip to Content
Merck
All Photos(1)

Key Documents

310867

Sigma-Aldrich

2-Fluoro-4-(trifluoromethyl)benzoic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€142.00

€142.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€142.00

About This Item

Linear Formula:
FC6H3(CF3)CO2H
CAS Number:
Molecular Weight:
208.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€142.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

168-170 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1ccc(cc1F)C(F)(F)F

InChI

1S/C8H4F4O2/c9-6-3-4(8(10,11)12)1-2-5(6)7(13)14/h1-3H,(H,13,14)

InChI key

OCIYTBZXTFPSPI-UHFFFAOYSA-N

General description

The molecular properties (both steric as well as electronic) of 2-fluoro-4-(trifluoromethyl)benzoic acid has been studied using molecular orbital and empirical methods.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Y Ghauri et al.
Biochemical pharmacology, 44(10), 1935-1946 (1992-11-17)
An extensive set of computed molecular properties, both steric and electronic, have been calculated using molecular orbital and empirical methods for benzoic acid (1) and a congeneric series of substituted benzoic acids, i.e. 2-, 3- and 4-fluorobenzoic acids (2-4), 2-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service