Skip to Content
Merck
All Photos(1)

Key Documents

309931

Sigma-Aldrich

Methyl palmitate

97%

Synonym(s):

n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2CH3
CAS Number:
Molecular Weight:
270.45
Beilstein:
1780973
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

97%

refractive index

n20/D 1.4512 (lit.)

bp

185 °C/10 mmHg (lit.)

mp

32-35 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3

InChI key

FLIACVVOZYBSBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tsutomu Ishioka et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(4), 671-680 (2003-03-01)
Normal mode analyses are made for methyl palmitate molecule having all-trans or conformational disorders around the ester head group, in order to explain characteristic observed frequency shifts in the wagging progressive region between all-trans and disorder chains in triglyceride molecules.
Ebtehal El-Demerdash
Toxicology and applied pharmacology, 254(3), 238-244 (2011-05-18)
Methyl palmitate (MP) has been shown earlier to inhibit Kupffer cells and rat peritoneal macrophages. To evaluate the potential of MP to inhibit the activation of other macrophages, RAW cells (macrophages of alveolar origin) were treated with varying concentrations of
Ari Gargir et al.
Biochimica et biophysica acta, 1569(1-3), 167-173 (2002-02-21)
The biological activities of many acylated molecules are lipid dependent. Lipids, however, are poorly immunogenic or non-immunogenic. We employed a phage display semi-synthetic human antibody library to isolate anti-lipid antibodies. Selection was done against methyl palmitate, a 16 carbon aliphatic
Eman M Mantawy et al.
Toxicology and applied pharmacology, 258(1), 134-144 (2011-11-15)
Fibrosis accompanies most chronic liver disorders and is a major factor contributing to hepatic failure. Therefore, the need for an effective treatment is evident. The present study was designed to assess the potential antifibrotic effect of MP and whether MP
Y N Wang et al.
Journal of economic entomology, 102(1), 196-202 (2009-03-04)
Walnut, Juglans regia L., is known for its insecticidal activities to a range of herbivores and microbes. Isolation and identification of bioactive compounds from walnut is a potential approach for the development of new pesticides. Laboratory experiments were carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service