Skip to Content
Merck
All Photos(1)

Key Documents

309311

Sigma-Aldrich

Mercury(II) perchlorate hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(ClO4)2·xH2O
CAS Number:
Molecular Weight:
399.49 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

composition

Degree of hydration, 3-5
Hg, 42-45%

reaction suitability

reagent type: oxidant

storage temp.

−20°C

SMILES string

O.[Hg++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Hg.H2O/c2*2-1(3,4)5;;/h2*(H,2,3,4,5);;1H2/q;;+2;/p-2

InChI key

OOYSCVKOXPZTGW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 3 - STOT RE 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service