Skip to Content
Merck
All Photos(1)

Key Documents

30750

Sigma-Aldrich

Dehydroacetic acid sodium salt

≥98.0% (NT)

Synonym(s):

2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid δ-lactone sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NaO4
CAS Number:
Molecular Weight:
190.13
Beilstein:
4893653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

mp

~295 °C (dec.)

SMILES string

[Na+].CC(=O)C1=C([O-])C=C(C)OC1=O

InChI

1S/C8H8O4.Na/c1-4-3-6(10)7(5(2)9)8(11)12-4;/h3,10H,1-2H3;/q;+1/p-1

InChI key

ZPNRBQVNNIDJHX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The kinetics of dehydroacetic acid sodium salt decomposition, in presence of formaldehyde and formaldehyde donors, was studied.

Application

Dehydroacetic acid sodium salt was used as a model preservative to study the relevant kinetic and formulation parameters of the device intended for the prolonged release of antimicrobial agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gazzaniga et al.
International journal of cosmetic science, 16(3), 105-112 (1994-06-01)
Synopsis The kinetics of decomposition of the sodium salt of dehydroacetic acid (DHA.Na) in presence of formaldehyde and formaldehyde donors was investigated. The possibility of preparing systems capable of providing the cosmetic preparation with increasing amounts of DHA.Na corresponding to
T Hata et al.
British journal of obstetrics and gynaecology, 105(3), 360-361 (1998-04-09)
We studied the effect of dehydroepiandrosterone sulphate (DHAS) on placental blood flow in 11 women with an uncomplicated pregnancy between 37 and 39 weeks of gestation. Power Doppler enhancement of the placenta was performed before and 60 minutes after the
Luiz C Dias et al.
Journal of agricultural and food chemistry, 57(4), 1399-1405 (2009-01-28)
A short and efficient approach to a range of new chiral and achiral functionalized (E)-enaminopyran-2,4-diones starting with commercially available dehydroacetic acid is described. The phytotoxic properties of these (E)-enaminopyran-2,4-diones were evaluated by their ability to interfere with the growth of
Allergic contact dermatitis caused by sodium dehydroacetate, not hyaluronic acid, in Ialuset® cream.
Brigitte Milpied et al.
Contact dermatitis, 65(6), 359-361 (2011-11-15)
Sodium dehydroacetate: an emerging allergen.
Caterina Foti et al.
Dermatitis : contact, atopic, occupational, drug, 23(5), 243-243 (2012-09-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service