Skip to Content
Merck
All Photos(2)

Key Documents

305499

Sigma-Aldrich

Sodium hexafluoroaluminate

97%

Synonym(s):

Cryolite, Cryolite, synthetic, Sodium aluminum hexafluoride

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€111.00

€111.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€111.00

About This Item

Linear Formula:
Na3AlF6
CAS Number:
Molecular Weight:
209.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

€111.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: aluminum

density

2.9 g/mL at 25 °C (lit.)

SMILES string

F[Al](F)(F)(F)(F)(F)([Na])([Na])[Na]

InChI

1S/Al.6FH.3Na/h;6*1H;;;/q+3;;;;;;;3*+1/p-6

InChI key

REHXRBDMVPYGJX-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Lact. - STOT RE 1 Oral

Target Organs

Lungs,Skeleton

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayanti Podder et al.
Toxicology and industrial health, 31(12), 1341-1347 (2013-07-13)
The study explored variations in the life cycle parameters in Drosophila melanogaster as a function of treatment with fluorinated insecticide, cryolite. Some of the life cycle parameters considered in this study were larval duration, pupal duration, and percentage of adult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service