Skip to Content
Merck
All Photos(2)

Key Documents

305111

Sigma-Aldrich

4-Chlorophenyl 2-bromoethyl ether

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€47.50

€47.50


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
€47.50

About This Item

Linear Formula:
ClC6H4OCH2CH2Br
CAS Number:
Molecular Weight:
235.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€47.50


Estimated to ship on13 April 2025


Request a Bulk Order

Assay

98%

form

solid

bp

165-170 °C/22 mmHg (lit.)

mp

40-42 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1ccc(OCCBr)cc1

InChI

1S/C8H8BrClO/c9-5-6-11-8-3-1-7(10)2-4-8/h1-4H,5-6H2

InChI key

YYFLBDSMQRWARK-UHFFFAOYSA-N

Application

4-Chlorophenyl 2-bromoethyl ether may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory J Baker et al.
Cell systems, 11(3), 272-285 (2020-09-09)
Accurately profiling systemic immune responses to cancer initiation and progression is necessary for understanding tumor surveillance and, ultimately, improving therapy. Here, we describe the SYLARAS software tool (systemic lymphoid architecture response assessment) and a dataset collected with SYLARAS that describes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service