Skip to Content
Merck
All Photos(1)

Key Documents

304093

Sigma-Aldrich

1,4-Dibromo-2-fluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3F
CAS Number:
Molecular Weight:
253.89
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

216 °C (lit.)

mp

33-36 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1cc(Br)ccc1Br

InChI

1S/C6H3Br2F/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

WNSNPGHNIJOOPM-UHFFFAOYSA-N

General description

The Suzuki-Miyaura reaction of 1,4-dibromo-2-fluorobenzene with arylboronic acids yields fluorinated para-terphenyls.[1]

Application

1,4-Dibromo-2-fluorobenzene was used in preparation of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Levitus et al.
The Journal of organic chemistry, 66(9), 3188-3195 (2001-04-28)
Crystals of 1,4-bis(2-hydroxy-2-methyl-3-butynyl)-2-fluorobenzene 4 have a rich packing structure with four distinct molecules in the unit cell. A complex hydrogen bonding network results in the formation of cofacial trimers, cofacial dimers, and monomers within the same unit cell. Given a
One-pot synthesis of fluorinated terphenyls by site-selective Suzuki-Miyaura reactions of 1, 4-dibromo-2-fluorobenzene.
Sharif M, et al.
Tetrahedron Letters, 51(21), 2810-2812 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service