All Photos(1)
About This Item
Linear Formula:
CH3CH=C(CH3)2
CAS Number:
Molecular Weight:
70.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
2.4 (vs air)
vapor pressure
14.34 psi ( 55 °C)
3.96 psi ( 20 °C)
form
liquid
concentration
2.0 M in THF
density
0.835 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
C\C=C(\C)C
InChI
1S/C5H10/c1-4-5(2)3/h4H,1-3H3
InChI key
BKOOMYPCSUNDGP-UHFFFAOYSA-N
General description
2-Methyl-2-butene acts as guest and forms stable solid host-guest complexes with self-assembled benzophenone bis-urea macrocycles. The impact of active chlorine on photooxidation of 2-methyl-2-butene was studied. Photosensitized oxidation of 2-methyl-2-butene adsorbed on internal framework of Na-ZSM-5 zeolite was studied.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
1.4 °F
Flash Point(C)
-17 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yunseok Im et al.
The Science of the total environment, 409(13), 2652-2661 (2011-04-29)
Active chlorine comprising hypochlorite (OCl⁻), hypochlorous acid (HOCl) and chlorine (Cl₂) is the active constituent in bleach formulations for a variety of industrial and consumer applications. However, the strong oxidative reactivity of active chlorine can cause adverse effects on both
Michael F Geer et al.
The Journal of organic chemistry, 78(11), 5568-5578 (2013-05-16)
This manuscript investigates how incorporation of benzophenone, a well-known triplet sensitizer, within a bis-urea macrocycle, which self-assembles into a columnar host, influences its photophysical properties and affects the reactivity of bound guest molecules. We further report the generation of a
Charles K Westbrook et al.
The journal of physical chemistry. A, 119(28), 7462-7480 (2015-03-31)
Two experimental studies have been carried out on the oxidation of 2-methyl-2-butene, one measuring ignition delay times behind reflected shock waves in a stainless steel shock tube, and the other measuring fuel, intermediate, and product species mole fractions in a
Confined space-controlled hydroperoxidation of trisubstituted alkenes adsorbed on pentasil zeolites.
Yu-Zhe Chen et al.
The Journal of organic chemistry, 70(12), 4676-4681 (2005-06-04)
Photosensitized oxidation of trialkylalkenes 2-methyl-2-pentene (1), 1-methylcyclohexene (2), trans-3-methyl-2-pentene (3), cis-3-methyl-2-pentene (4), and 2-methyl-2-butene (5) included in the internal framework of Na-ZSM-5 zeolites was investigated. The zeolite samples having adsorbed the alkenes were suspended in isooctane, and the sensitizer, tetraphenylporphyrin
Masataka Ikeda et al.
Scientific reports, 5, 15881-15881 (2015-10-31)
The heart has two major modalities of hypertrophy in response to hemodynamic loads: concentric and eccentric hypertrophy caused by pressure and volume overload (VO), respectively. However, the molecular mechanism of eccentric hypertrophy remains poorly understood. Here we demonstrate that the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service