Skip to Content
Merck
All Photos(1)

Key Documents

303933

Sigma-Aldrich

3-Butylpyridine

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

80-81 °C/8 mmHg (lit.)

density

0.911 g/mL at 25 °C (lit.)

SMILES string

CCCCc1cccnc1

InChI

1S/C9H13N/c1-2-3-5-9-6-4-7-10-8-9/h4,6-8H,2-3,5H2,1H3

InChI key

QSNMFWFDOFQASV-UHFFFAOYSA-N

General description

3-Butylpyridine is major component of volatile oil isolated from aerial parts of Pavonia odorata.[1]

Application

3-Butylpyridine was used in preparation of series of manganese-salen (salen = N,N′-bis(salicylidene)ethylenediamine) complexes.[2] It was also used as starting material for the synthesis of 3-butylisonicotinic acid.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yusei Kashima et al.
Journal of oleo science, 63(2), 149-158 (2014-01-21)
The chemical composition of volatile oil obtained from aerial parts of Pavonia odorata were investigated using gas chromatography-mass spectrometry (GC-MS). Its aroma-active compounds were identified using gas chromatography-olfactometry (GC-O) and aroma extraction dilution analysis (AEDA). In order to determine the
Studies in the pyridine series. XIV. Novel procedures of preparing 3-butylisonicotinic acid.
Ferles M and Jankovsky M.
Collection of Czechoslovak Chemical Communications, 31(7), 3008-3011 (1966)
Thermal properties and crystal structures of manganese (III)-salen complexes with the Tf2 N anion [Tf2 N= bis (trifluoromethanesulfonyl) amide)].
Okuhata M and Mochida T.
Polyhedron, 43(1), 153-158 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service