Skip to Content
Merck
All Photos(2)

Key Documents

303550

Sigma-Aldrich

3,5,5-Trimethylhexanal

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€78.90

€78.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€78.90

About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
142.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€78.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

67-68 °C/2.5 mmHg (lit.)

density

0.817 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CC(CC=O)CC(C)(C)C

InChI

1S/C9H18O/c1-8(5-6-10)7-9(2,3)4/h6,8H,5,7H2,1-4H3

InChI key

WTPYRCJDOZVZON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5,5-Trimethylhexanal was used in preparation of gem-dithiols via reaction with hydrogen sulfide.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

gem-Dithiols1.
Cairns TL, et al.
Journal of the American Chemical Society, 74(16), 3982-3989 (1952)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service