Skip to Content
Merck
All Photos(1)

Key Documents

302759

Sigma-Aldrich

Benzylmagnesium chloride solution

1.0 M in diethyl ether

Synonym(s):

(Phenylmethyl)magnesium chloride, Benzylchloromagnesium, Chloro(phenylmethyl)magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€98.10
800 ML
€363.00

€98.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€98.10
800 ML
€363.00

About This Item

Linear Formula:
C6H5CH2MgCl
CAS Number:
Molecular Weight:
150.89
Beilstein:
878506
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€98.10


Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.794 g/mL at 25 °C

functional group

phenyl

SMILES string

Cl[Mg]Cc1ccccc1

InChI

1S/C7H7.ClH.Mg/c1-7-5-3-2-4-6-7;;/h2-6H,1H2;1H;/q;;+1/p-1

InChI key

NRAFPLGJPPJUNB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Benzylmagnesium chloride is a versatile Grignard reagent frequently used in organic synthesis. It can be used as a reagent in the addition reaction with carbonyl compounds, nitriles, epoxides, and alkyl halides to synthesize various alcohols, amines, and ketones. Additionally, it is also used in reductions, cross-coupling, allylation and alkylation reactions.

Application

Benzylmagnesium chloride solution (1.0 M in diethyl ether) is used as a reagent to provide the benzyl group required for the synthesis of the desired sulfonic acid-functionalized alkoxysilane. It is also used in the anionic polymerization of styrene to form styrenyl magnesium chloride, which is further used as an initiator in the synthesis of polyolefin-block-polystyrene block copolymers.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service