Skip to Content
Merck
All Photos(2)

Key Documents

302384

Sigma-Aldrich

2,4,4,6-Tetrabromo-2,5-cyclohexadienone

90%

Synonym(s):

TBCO

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€68.60

€68.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€68.60

About This Item

Empirical Formula (Hill Notation):
C6H2Br4O
CAS Number:
Molecular Weight:
409.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€68.60


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

90%

form

solid

functional group

bromo
ketone

storage temp.

2-8°C

SMILES string

BrC1=CC(Br)(Br)C=C(Br)C1=O

InChI

1S/C6H2Br4O/c7-3-1-6(9,10)2-4(8)5(3)11/h1-2H

InChI key

NJQJGRGGIUNVAB-UHFFFAOYSA-N

General description

2,4,4,6-Tetrabromo-2,5-cyclohexadienone forms solid charge-transfer molecular complexes with 4-(aminomethyl)piperidine and their spectroscopic characteristics were studied.[1]

Application

2,4,4,6-Tetrabromo-2,5-cyclohexadienone was used as catalyst for efficient deoxygenation of sulfoxides to their corresponding sulfides with 1,3-dithiane.[2] It was also used as reagent for conversion of alcohols to azides.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jakob Gustavsson et al.
Journal of chromatography. A, 1524, 74-86 (2017-10-17)
The influence of natural organic matter (NOM) on the solid-phase extraction (SPE) efficiency was investigated for legacy and emerging flame retardants (FRs; n=26) in surface water. Three different groups of FRs were analyzed: polybrominated diphenyl ethers (PBDEs), halogenated flame retardants
Darren Van Essen et al.
Chemosphere, 266, 129195-129195 (2020-12-15)
Brominated flame retardants (BFRs) can enter aquatic environments where they can have adverse effects on organisms. The BFR, 1,2,5,6-Tetrabromocyclooctane (TBCO), has been introduced as a potential replacement for the major use BRF, Hexabromocyclododecane (HBCD). However, little is known about effects
Adel Mostafa et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 118, 1012-1019 (2013-11-05)
The spectroscopic characteristics of the solid charge-transfer molecular complexes (CT) formed in the reaction of the electron donor 4-(aminomethyl) piperidine (4AMP) with the σ-acceptor iodine and the π-acceptors 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ), 2,4,4,6-tetrabromo-2,5-cyclohexadienone (TBCHD) and 7,7,8,8-tetracyanoquinodimethane (TCNQ) have been studied in chloroform
An efficient method for converting alcohols to azides with 2, 4, 4, 6-tetrabromo-2, 5-cyclohexadienone/PPh3 /Zn (N3)2? 2Py.
Saito A, et al.
Tetrahedron Letters, 38(22), 3955-3958 (1997)
Giulia Simonetti et al.
International journal of environmental research and public health, 17(11) (2020-05-31)
The occurrence of halogenated organic pollutants in indoor dust can be high due to the presence of textile, electronic devices, furniture, and building materials treated with these chemicals. In this explorative study, we focused on emerging organic pollutants, such as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service