Skip to Content
Merck
All Photos(1)

Key Documents

302104

Sigma-Aldrich

n-Butyllithium solution

2.0 M in pentane

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
1209227
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

concentration

2.0 M in pentane

density

0.658 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yuqiang Ma et al.
ACS nano, 9(7), 7383-7391 (2015-07-01)
Two-dimensional (2D) semiconducting monolayer transition metal dichalcogenides (TMDCs) have stimulated lots of interest because they are direct bandgap materials that have reasonably good mobility values. However, contact between most metals and semiconducting TMDCs like 2H phase WSe2 are highly resistive
Karl J Hale et al.
Organic letters, 5(4), 503-505 (2003-02-14)
[reaction: see text] The "Southern Hemisphere" intermediate 2, used by Masamune and co-workers for their asymmetric total synthesis of bryostatin 7 (1), has been synthesized from (E)-1,4-hexadiene (11) by a 24-step pathway that has a longest linear sequence of only
Jennifer L Rutherford et al.
Journal of the American Chemical Society, 124(2), 264-271 (2002-01-10)
6Li and (13)C NMR spectroscopic studies were carried out on [(6)Li]n-BuLi and [(6)Li]PhLi (RLi) in toluene-d(8) containing the following diamines: N,N,N',N'-tetramethylethylenediamine (TMEDA), N,N,N',N'-tetraethylethylenediamine, 1,2-dipyrrolidinoethane, 1,2-dipiperidinoethane, N,N,N',N'-tetramethylpropanediamine, trans-(R,R)-N,N,N',N'-tetramethylcyclohexanediamine, and (-)-sparteine. Dimers of general structure (RLi)(2)S(2) (S = chelating diamine) are formed
Enantioselective total synthesis of the marine toxin (-)-gymnodimine employing a Barbier-type macrocyclization.
Ke Kong et al.
Angewandte Chemie (International ed. in English), 48(40), 7402-7405 (2009-09-04)
Deyu Li et al.
The Journal of organic chemistry, 73(6), 2373-2381 (2008-02-26)
A system consisting of a chiral lithium amide and n-BuLi in tol-d(8) solution was investigated with (1)H and (13)C INEPT DOSY, (6)Li and (15)N NMR, and other 2D NMR techniques. A mixed 2:1 trimeric complex was identified as the major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service