Skip to Content
Merck
All Photos(3)

Key Documents

302066

Sigma-Aldrich

1,2-Dinitrobenzene

≥99%

Synonym(s):

o-Dinitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NO2)2
CAS Number:
Molecular Weight:
168.11
Beilstein:
642224
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

solid

bp

319 °C/773 mmHg (lit.)

mp

114-117 °C (lit.)

solubility

chloroform: soluble 5%, clear, yellow-green

SMILES string

[O-][N+](=O)c1ccccc1[N+]([O-])=O

InChI

1S/C6H4N2O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H

InChI key

IZUKQUVSCNEFMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The electrochemistry of 1,2-dinitrobenzene in the presence of 1,3-diphenylurea was studied.[1] The kinetics of the aromatic nucleophilic substitution reactions of 1,2-dinitrobenzene with butylamine and piperidine were investigated as a function of the amine concentration and temperature.[2]

Application

1,2-Dinitrobenzene was used as internal standard for analysis of the explosives, TNT, RDX, and tetryl in sea water by vapor phase chromatography with the nickel-63 electron capture detector.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative kinetic study of solvent effects in the reactions of 1, 2-dinitrobenzene with butylamine and piperidine.
Chiacchiera SM, et al.
Journal of Physical Organic Chemistry, 2(8), 631-645 (1989)
Analysis of explosives in sea water.
Hoffsommer JC and Rosen JM.
Bulletin of Environmental Contamination and Toxicology, 7(2-3), 177-181 (1972)
Cindy Chan-Leonor et al.
The Journal of organic chemistry, 70(26), 10817-10822 (2005-12-17)
[reaction: see text] The electrochemistry of 1,2-dinitrobenzene (1,2-DNB), 1,3-dinitrobenzene (1,3-DNB), and 1,4-dinitrobenzene (1,4-DNB) is strongly affected by the presence of 1,3-diphenylurea. In DMF, the second reduction potential of all three DNBs shifts substantially positive in the presence of the urea
J V Barbo et al.
Journal of immunology (Baltimore, Md. : 1950), 154(4), 1551-1559 (1995-02-15)
A population of CD8+ T cells from dinitrobenzene sulfonate-primed mice produce soluble effector molecules that down-regulate the magnitude of dinitrophenol-specific contact hypersensitivity reactions. These soluble molecules express the binding specificity and serologic determinants of alpha/beta TCR. To examine the requirement
H Tamaki et al.
Journal of bacteriology, 181(9), 2958-2962 (1999-04-28)
Glutathione S-transferases (GSTs) Y-1 and Y-2 from the yeast Issatchenkia orientalis were purified by passage through a glutathione-agarose column, and the cDNA for GST Y-1 was cloned and sequenced. The deduced amino acid sequence consisted of 188 residues with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service