Skip to Content
Merck
All Photos(2)

Key Documents

301620

Sigma-Aldrich

3-Bromo-2-(bromomethyl)propionic acid

97%

Synonym(s):

β,β′-Dibromoisobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(BrCH2)2CHCO2H
CAS Number:
Molecular Weight:
245.90
Beilstein:
1752505
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

mp

98-101 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

OC(=O)C(CBr)CBr

InChI

1S/C4H6Br2O2/c5-1-3(2-6)4(7)8/h3H,1-2H2,(H,7,8)

InChI key

QQZJWQCLWOQDQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Bromo-2-(bromomethyl)propionic acid reacts with alkaline arsenite to yield (RS)-3-arsono-2-(hydroxymethyl)propionic acid.[1]

Application

3-Bromo-2-(bromomethyl)propionic acid was used in the synthesis of t-butyl 2-(phenylthiomethyl)propenoate, t-butyl and methyl 3-(phenylthio)-2-(phenylthiomethyl)propenoate and 3-(phenylthio)-2-(phenyl- sulfinylmethyl)propenoate.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Chawla et al.
Journal of enzyme inhibition, 8(4), 255-259 (1995-01-01)
(RS)-3-Arsono-2-(hydroxymethyl)propionic acid was synthesized by the action of alkaline arsenite on 3-bromo-2-(bromomethyl)propionic acid. It is a substrate for yeast enolase (EC 4.2.1.11) with a Km of 6.5 mM (for 2-phospho-D-glycerate Km = 0.08 mM). The catalytic constant of the enzyme
Preparation of t-Butyl 2-(Phenylthiomethyl) propenoate, t-Butyl 3-(Phenylthio)-2-(phenylthiomethyl) propenoate and related compounds.
Haynes RK, et al.
Australian Journal of Chemistry, 37(7), 1571-1578 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service