Skip to Content
Merck
All Photos(1)

Key Documents

301167

Sigma-Aldrich

4′-Cyclohexylacetophenone

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€73.70

€73.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€73.70

About This Item

Linear Formula:
C6H11C6H4COCH3
CAS Number:
Molecular Weight:
202.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€73.70


Check Cart for Availability

Request a Bulk Order

Assay

99%

bp

179-181 °C/14 mmHg (lit.)

mp

68-70 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1ccc(cc1)C2CCCCC2

InChI

1S/C14H18O/c1-11(15)12-7-9-14(10-8-12)13-5-3-2-4-6-13/h7-10,13H,2-6H2,1H3

InChI key

MSDQNIRGPBARGC-UHFFFAOYSA-N

General description

Antimycobacterial activity of 4-cyclohexylacetophenone was studied.[1] The tricarbonylchromium complexes of 4-cyclohexylacetophenone were prepared and spectral properties were studied.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of restricted rotation on the spectral properties of acetophenonetricarbonylchromium complexes.
Armstrong BM, et al.
Inorgorganica Chimica Acta, 170(1), 37-43 (1990)
L Rajabi et al.
Letters in applied microbiology, 40(3), 212-217 (2005-02-18)
Mycobacteria are a serious cause of infections in humans, with limited treatment options, as no new antibiotics have been developed against mycobacteria since the 1960s. In this study, the antimycobacterial activity of a small library of acetophenone (AP) compounds was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service