300969
5-Bromo-1,2,3,4,5-pentaphenyl-1,3-cyclopentadiene
technical grade, 90%
About This Item
grade
technical grade
Assay
90%
form
solid
mp
188-190 °C (lit.)
functional group
bromo
phenyl
SMILES string
BrC2(c1ccccc1)C(c3ccccc3)=C(c4ccccc4)C(c5ccccc5)=C2c6ccccc6
InChI
1S/C35H25Br/c36-35(30-24-14-5-15-25-30)33(28-20-10-3-11-21-28)31(26-16-6-1-7-17-26)32(27-18-8-2-9-19-27)34(35)29-22-12-4-13-23-29/h1-25H
InChI key
HCSNYWQHJSONEK-UHFFFAOYSA-N
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service