Skip to Content
Merck
All Photos(2)

Key Documents

300780

Sigma-Aldrich

Ethyl 4-pyrazolecarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

bp

138-140 °C/3 mmHg (lit.)

mp

78-80 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cn[nH]c1

InChI

1S/C6H8N2O2/c1-2-10-6(9)5-3-7-8-4-5/h3-4H,2H2,1H3,(H,7,8)

InChI key

KACZQOKEFKFNDB-UHFFFAOYSA-N

Application

Ethyl 4-pyrazolecarboxylate was used in the preparation of 4-chloromethylpyrazole.[1] It was also used as starting material in a copper-diamine-catalyzed N-arylation reaction.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jon C Antilla et al.
The Journal of organic chemistry, 69(17), 5578-5587 (2004-08-17)
This paper details the copper-catalyzed N-arylation of pi-excessive nitrogen heterocycles. The coupling of either aryl iodides or aryl bromides with common nitrogen heterocycles (pyrroles, pyrazoles, indazoles, imidazoles, and triazoles) was successfully performed in good yield with catalysts derived from diamine
J F Schindler et al.
Journal of protein chemistry, 15(8), 737-742 (1996-11-01)
Improved and efficient techniques have led to an explosive growth in the application of site-directed mutagenesis to the study of enzymes. However, the limited availability of only those 20 amino acids that are translated by the genetic code has prevented

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service