Skip to Content
Merck
All Photos(1)

Documents

300004

Sigma-Aldrich

Trifluoroacetyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COCl
CAS Number:
Molecular Weight:
132.47
Beilstein:
1098994
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.6 (vs air)

Quality Level

Assay

98%

bp

−27 °C (lit.)

mp

−146 °C (lit.)

SMILES string

FC(F)(F)C(Cl)=O

InChI

1S/C2ClF3O/c3-1(7)2(4,5)6

InChI key

PNQBEPDZQUOCNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a 316 stainless steel diaphragm valve with a male 1/4" NPTF outlet thread (Z168637) installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service