Skip to Content
Merck
All Photos(2)

Key Documents

298069

Sigma-Aldrich

5-Decanolide

≥98%

Synonym(s):

(±)-5-Decanolide, δ-Decanolactone, (±)-δ-Pentyl-δ-valerolactone, (±)-6-Pentyltetrahydro-2H-pyran-2-one, 5-Hydroxydecanoic acid δ-lactone

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€105.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€105.00

About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Beilstein:
117520
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€105.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.458 (lit.)

bp

117-120 °C/0.02 mmHg (lit.)

mp

−27 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CCCCCC1CCCC(=O)O1

InChI

1S/C10H18O2/c1-2-3-4-6-9-7-5-8-10(11)12-9/h9H,2-8H2,1H3

InChI key

GHBSPIPJMLAMEP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Measurement of optical rotatory dispersion and circular dichroism of (+) 5-decanolide (tetrahydro-6-pentyl-2H-pyran-2-one) has been reported[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical rotatory dispersion and circular dichroism of d-lactones: Determination of the absolute configuration of (+) 5-decanolide and (+) 5-dodecanolide.
Korver O.
Tetrahedron, 26(10), 2391-2396 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service