Skip to Content
Merck
All Photos(1)

Documents

298042

Sigma-Aldrich

Sodium acetate-1-13C,d3

99 atom % 13C, 99 atom % D

Synonym(s):

Acetic acid-1-13C-2,2,2-d3 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD313CO2Na
CAS Number:
Molecular Weight:
86.04
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C
99 atom % D

Quality Level

form

solid

mp

>300 °C (dec.) (lit.)

mass shift

M+4

SMILES string

[Na+].[2H]C([2H])([2H])[13C]([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1D3,2+1;

InChI key

VMHLLURERBWHNL-CKHMYQJJSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petia Kovatcheva-Datchary et al.
Cell reports, 26(13), 3772-3783 (2019-03-28)
The gut microbiota can modulate human metabolism through interactions with macronutrients. However, microbiota-diet-host interactions are difficult to study because bacteria interact in complex food webs in concert with the host, and many of the bacteria are not yet characterized. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service