Skip to Content
Merck
All Photos(2)

Key Documents

295817

Sigma-Aldrich

(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene

97%

Synonym(s):

(R)-(+)-(1,1′-Binaphthalene-2,2′-diyl)bis(diphenylphosphine), (R)-(+)-BINAP

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€80.50
5 G
€124.00

€80.50


Estimated to ship on31 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
€80.50
5 G
€124.00

About This Item

Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€80.50


Estimated to ship on31 March 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

optical activity

[α]19/D +233°, c = 0.3 in toluene

optical purity

ee: 99% (HPLC)

mp

239-241 °C (lit.)

functional group

phosphine

SMILES string

c1ccc(cc1)P(c2ccccc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccccc7)c8ccccc8

InChI

1S/C44H32P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32H

InChI key

MUALRAIOVNYAIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-BINAP is a chiral diphosphine ligand that exhibits high enantioselectivity and reactivity in various organic reactions.[1]

(R)-(+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene is an axially dissymmetric bis(triaryl)phosphine ligand for asymmetric reactions.[2]

Application

Useful ligand for transition metal catalyzed asymmetric reactions, including hydrogenation[3] and disilylation.[4] 2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl and its rhodium and ruthenium derivatives are highly selective homogeneous catalysts used for the reduction of aryl ketones,[5] β-keto esters,[6] and α-amino ketones.[7] They have also been used for asymmetric hydrogenation[8] and hydroformylation of olefins,[9] asymmetric Heck reactions,[10] and asymmetric isomerizations of allyls.[11][12]
Complex with Ag(I) used to catalyze an asymmetric aldol reaction between alkenyl trichloroacetates and aldehydes.[13] Also used with Ag(I) to catalyze an enantioselective hetero-Diels-Alder reaction of azo compounds.[14]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 71, 1-1 (1993)
Tetrahedron, 50, 335-335 (1994)
Masanori Kawasaki et al.
Journal of the American Chemical Society, 128(51), 16482-16483 (2006-12-21)
This communication describes studies in which an azo hetero-Diels-Alder adduct was furnished in high regio- and enantioselectivity using azopyridine as a reagent and silver as a catalyst. The obtained hetero-Diels-Alder adduct was easily converted to the corresponding chiral 1,4-diamino alcohol.
Journal of Organometallic Chemistry, 692, 550-550 (2007)
(R)-& (S)-2, 2?-Bis (diphenylphosphino)-1, 1?-binaphthyl
Kitamura M, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)

Articles

We present an article concerning BINAP/SEGPHOS® Ligands and Complexes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service