Skip to Content
Merck
All Photos(1)

Key Documents

294594

Sigma-Aldrich

Mercury

electronic grade, 99.9999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hg
CAS Number:
Molecular Weight:
200.59
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

electronic grade

vapor density

7 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 126 °C)

Assay

99.9999% trace metals basis

form

liquid

resistivity

95.8 μΩ-cm, 20°C

bp

356.6 °C (lit.)

mp

−38.87 °C (lit.)

SMILES string

[Hg]

InChI

1S/Hg

InChI key

QSHDDOUJBYECFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ekim Seven et al.
Journal of hypertension, 32(7), 1488-1494 (2014-05-09)
The adipocytokines, leptin, adiponectin, and interleukin-6, which stimulate liver C-reactive protein (CRP) production, are regarded as potential candidate intermediates between adipose tissue and overweight-induced hypertension. We examined the associations between leptin, adiponectin, and CRP levels with both prevalent and 5-year
Okechukwu S Ogah et al.
JACC. Heart failure, 2(3), 250-259 (2014-06-24)
The aim of this study was to determine the contemporary profile, clinical characteristics, and intrahospital outcomes of acute heart failure (AHF) in an African urban community. There are limited data on the current burden and characteristics of AHF in Nigerian
Sabrina Llop et al.
PloS one, 9(5), e97172-e97172 (2014-05-17)
The genetic background may influence methylmercury (MeHg) metabolism and neurotoxicity. ATP binding cassette (ABC) transporters actively transport various xenobiotics across biological membranes. To investigate the role of ABC polymorphisms as modifiers of prenatal exposure to MeHg. The study population consisted
M E Goodsite et al.
The Science of the total environment, 452-453, 196-207 (2013-03-20)
This review compares the reconstruction of atmospheric Hg deposition rates and historical trends over recent decades in the Arctic, inferred from Hg profiles in natural archives such as lake and marine sediments, peat bogs and glacial firn (permanent snowpack), against
James P K Rooney
Toxicology and applied pharmacology, 274(3), 425-435 (2013-12-26)
Reports from human case studies indicate a half-life for inorganic mercury in the brain in the order of years-contradicting older radioisotope studies that estimated half-lives in the order of weeks to months in duration. This study systematically reviews available evidence

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. I see that Mercury is sold by weight, but it's a liquid. What is the volume?

    1 answer
    1. The specific gravity of liquid mercury at 20°C is 13.546, according to the CRC Handbook of Chemistry and Physics, 82nd ed. (2001-2002), page 4-19. We list several mercury products, in various package sizes, sold by weight. Here are the conversions from mass to volume: 5G = 0.37 mL. 50G = 3.7 mL. 100G = 7.4 mL. 250G = 18.5 mL. 500G = 36.9 mL. 2KG = 147.6 mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service