Skip to Content
Merck
All Photos(1)

Documents

294071

Sigma-Aldrich

Methoxymethyl phenyl sulfide

97%

Synonym(s):

[(Methoxymethyl)thio]benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH2OCH3
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.5632 (lit.)

bp

113-114 °C/18 mmHg (lit.)

density

1.047 g/mL at 25 °C (lit.)

SMILES string

COCSc1ccccc1

InChI

1S/C8H10OS/c1-9-7-10-8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

QPXQVNXSQCRWEV-UHFFFAOYSA-N

General description

Meisenheimer rearrangement of methoxymethyl phenyl sulfoxide to methoxymethyl benzenesulfenate has been reported. Methoxymethyl phenyl sulfide undergoes solvent free permanganate oxidation to yield methoxymethyl phenyl sulfone.

Application

Methoxymethyl phenyl sulfide has been used in the preparation of methoxymethyl phenyl sulfoxide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent free permanganate oxidations.
Shaabani A and Lee DG.
Tetrahedron Letters, 42(34), 5833-5836 (2001)
Meisenheimer rearrangement of methoxymethyl phenyl sulfoxide. Formation and disproportionation of methoxymethyl benzenesulfenate.
Maricich TJ and Harrington CK.
Journal of the American Chemical Society, 94(14), 5115-5116 (1972)
S P Hanlon et al.
Microbiology (Reading, England), 144 ( Pt 8), 2247-2253 (1998-08-28)
The enantioselective reduction of racemic sulfoxides by dimethyl sulfoxide reductases from Rhodobacter capsulatus, Escherichia coli, Proteus mirabilis and Proteus vulgaris was investigated. Purified dimethyl sulfoxide reductase from Rhodobacter capsulatus catalysed the selective removal of (S)-methyl p-tolyl sulfoxide from a racemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service