Skip to Content
Merck
All Photos(1)

Key Documents

293008

Sigma-Aldrich

Trimethylsulfoxonium chloride

98%

Synonym(s):

NSC 221182

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€127.00

€127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€127.00

About This Item

Linear Formula:
(CH3)3S(Cl)O
CAS Number:
Molecular Weight:
128.62
Beilstein:
4549927
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€127.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

226-229 °C (lit.)

functional group

sulfoxide

SMILES string

[Cl-].C[S+](C)(C)=O

InChI

1S/C3H9OS.ClH/c1-5(2,3)4;/h1-3H3;1H/q+1;/p-1

InChI key

KQYWHJICYXXDSQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to dimethyloxosulfonium methylide.
Reactant involved in:
  • Polymerization of ylides
  • X-H insertion
  • One-pot sequential addition
  • Cyclopropanation of enones

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service