Skip to Content
Merck
All Photos(1)

Key Documents

29295

Sigma-Aldrich

3-Cyclohexyl-D-alanine hydrate

≥99.0% (calc. based on dry substance, NT), ~1 mol/mol water

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2 · xH2O
CAS Number:
Molecular Weight:
171.24 (anhydrous basis)
Beilstein:
3197315
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (calc. based on dry substance, NT)

form

solid

optical activity

[α]20/D −12±1°, c = 1% in 1 M HCl (dry matter)

impurities

~1 mol/mol water

application(s)

peptide synthesis

SMILES string

O.N[C@H](CC1CCCCC1)C(O)=O

InChI

1S/C9H17NO2.H2O/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h7-8H,1-6,10H2,(H,11,12);1H2/t8-;/m1./s1

InChI key

HDJQIWAIDCEDEF-DDWIOCJRSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Aarstad et al.
European journal of biochemistry, 112(2), 335-338 (1980-11-01)
The amino acid analog L-cyclohexylalanine, which may be considered an analog of the hydrophobic amino acids leucine and valine, was found to thioester-bind to the heavy enzyme of gramicidin S synthetase. The results indicate that it preferably binds to the
Géza Tóth et al.
Journal of medicinal chemistry, 50(2), 328-333 (2007-01-19)
The opioid peptide TIPP (H-Tyr-Tic-Phe-Phe-OH, Tic:1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid) was substituted with Dmt (2',6'-dimethyltyrosine) and a new unnatural amino acid, beta-MeCha (beta-methyl-cyclohexylalanine). This double substitution led to a new series of opioid peptides displaying subnanomolar delta antagonist activity and mu agonist or
Jana Tchekalarova et al.
Brain research, 1032(1-2), 94-103 (2005-02-01)
The effects of the hexapeptide angiotensin II (3-8) ANG IV, the selective A(1) receptor agonist cyclohexyladenosine (CHA) and the combination of ANG IV + CHA on pentylenetetrazol (PTZ)-generalized seizures; kindling development and maintenance were studied. By using in vitro quantitative
Nathan A Schnarr et al.
Journal of the American Chemical Society, 125(3), 667-671 (2003-01-16)
Designed coiled-coil heterotrimers are described whose assembly is governed by both hydrophobic and hydrophilic forces. Sterically matched hydrophobic core side-chain packing of alanine and cyclohexylalanine has been shown to promote formation of a 1:1:1 heterotrimer. Manipulation of hydrophilic glutamic acid
J Hondrelis et al.
International journal of peptide and protein research, 37(1), 21-26 (1991-01-01)
Analogues of angiotensin II with cyclohexylalanine (Cha) at position 4 or 8, and analogues of the competitive (type II) angiotensin antagonist [Sar1,Tyr(Me)4]ANG II (Sarmesin) with Cha at position 8, have been prepared by the solid phase method and purified by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service