Skip to Content
Merck
All Photos(2)

Key Documents

287679

Sigma-Aldrich

Lithium trifluoroacetate

95%

Synonym(s):

Trifluoroacetic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2Li
CAS Number:
Molecular Weight:
119.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
form:
solid

Assay

95%

form

solid

functional group

fluoro

SMILES string

[Li+].[O-]C(=O)C(F)(F)F

InChI

1S/C2HF3O2.Li/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

HSFDLPWPRRSVSM-UHFFFAOYSA-M

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Shigenobu et al.
Physical chemistry chemical physics : PCCP, 23(4), 2622-2629 (2021-01-22)
To achieve single-ion conducting liquid electrolytes for the rapid charge and discharge of Li secondary batteries, improvement in the Li+ transference number of the electrolytes is integral. Few studies have established a feasible design for achieving Li+ transference numbers approaching
Lars Schmüser et al.
ACS applied materials & interfaces, 12(10), 12294-12304 (2020-02-11)
Collembola are ancient arthropods living in soil with extensive exposure to dirt, bacteria, and fungi. To protect from the harsh environmental conditions and to retain a layer of air for breathing when submerged in water, they have evolved a superhydrophobic
Ting Cheng et al.
Acta biomaterialia, 54, 186-200 (2017-03-21)
Near-infrared (NIR) triggered chemical delivery allows on-demand release with the advantage of external tissue stimulation. Bioresorbable polyester poly-l-lactic acid (PLLA) was compounded with photoadditives of neat zinc oxide (ZnO) nanoparticles and 980→365nm LiYF Upconverting nanoparticles and zinc oxide nanorods were
Ines Schreiver et al.
The Analyst, 143(16), 3941-3950 (2018-07-28)
With regard to the increasing number of tattooed people, legal regulations for tattoo inks were implemented across Europe-aiming for higher consumer safety. To control the laws' abidance, analytical methods are needed to identify banned ingredients from the given negative lists.
Tuhin Samanta et al.
Dalton transactions (Cambridge, England : 2003), 46(29), 9646-9653 (2017-07-15)
Lanthanide (Ln)-doped nanocrystals generally display low luminescence quantum efficiency due to forbidden nature of the 4f-4f transition besides possessing low absorption cross sections (∼10 M

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service