Skip to Content
Merck
All Photos(1)

Documents

287261

Sigma-Aldrich

Tetratriacontane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)32CH3
CAS Number:
Molecular Weight:
478.92
Beilstein:
1788466
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

flakes

bp

285 °C/2 mmHg (lit.)

mp

72-75 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C34H70/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-34H2,1-2H3

InChI key

GWVDBZWVFGFBCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetratriacontane is one of the components of the oil extracts of the red alga Jania ruben.

Application

Tetratriacontane was used as an internal standard in the analysis of a new estrogenic drug, 17 α-ethynylestriol 3-cyclopentyl ether, by GLC method.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R H Bishara et al.
Journal of pharmaceutical sciences, 64(7), 1210-1213 (1975-07-01)
A rapid, sensitive, and accurate GLC method of analysis of a new estrogenic drug, 17 alpha-ethynylestriol 3-cyclopentyl ether, was developed. The drug and the internal standard, tetratriacontane, are dissolved in chloroform, and an aliquot is heated with N-trimethylsilylimidazole at 80
N Ulku Karabay-Yavasoglu et al.
Phytotherapy research : PTR, 21(2), 153-156 (2006-11-28)
The methanol, dichloromethane, hexane, chloroform and volatile oil extracts of the red alga Jania rubens were tested in vitro for their antimicrobial activity (five Gram-positive, four Gram-negative bacteria and Candida albicans ATCC 10239). GC-MS analysis of the volatile components of
Johannes Niebler et al.
Phytochemistry, 109, 66-75 (2014-12-04)
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service