Skip to Content
Merck
All Photos(1)

Key Documents

283479

Sigma-Aldrich

Diethyl diallylmalonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCH2)2C(CO2C2H5)2
CAS Number:
Molecular Weight:
240.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

128-130 °C/12 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

functional group

allyl
ester

SMILES string

CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

InChI

1S/C13H20O4/c1-5-9-13(10-6-2,11(14)16-7-3)12(15)17-8-4/h5-6H,1-2,7-10H2,3-4H3

InChI key

LYUUVYQGUMRKOV-UHFFFAOYSA-N

General description

Low catalyst loading in ring-closing metathesis reaction of diethyl diallylmalonate has been reported[1]. Cationic nickel (with monodentate phosphoramidites and Wilke′s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Böing et al.
Chemical communications (Cambridge, England), (11)(11), 1456-1458 (2005-03-10)
Cationic nickel catalysts with monodentate phosphoramidites and Wilke's azaphospholene as ligands are highly regio- and enantioselective catalysts for the cycloisomerisation of diethyl diallylmalonate.
Renat Kadyrov
Chemistry (Weinheim an der Bergstrasse, Germany), 19(3), 1002-1012 (2012-11-28)
An efficient procedure is described for ring-closing metathesis reactions. A conversion of 95% for diethyl diallylmalonate in dilute solution could be achieved within a few minutes, reaching TOF = 4173 min(-1), with very low loading of commercially available Ru catalysts that

Articles

Effective in key synthesis reactions like Knoevenagel condensation, thalidomide synthesis, and Suzuki coupling for sustainable chemical transformations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service