Skip to Content
Merck
All Photos(1)

Key Documents

282634

Sigma-Aldrich

Tritriacontane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)31CH3
CAS Number:
Molecular Weight:
464.89
Beilstein:
1786920
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

71-73 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C33H68/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-33H2,1-2H3

InChI key

SUJUOAZFECLBOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Dorset et al.
Journal of electron microscopy technique, 18(2), 142-147 (1991-06-01)
The crystal structure of the odd-chain paraffin, n-tritriacontane, nC33H68, is determined directly by using low-dose electron microscope images and electron diffraction intensity data from epitaxially grown microcrystals. Phases of the most intense "polyethylene" reflections are determined from triplet structure-invariant relationships
Gang Wang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 33(4), 551-554 (2010-09-18)
To study the chemical constituents from Kalimeris indica. Compounds were isolated and purified with silica gel and Sephadex LH-20 column chromatography, their structures were determined by using spectroscopic analysis including MS and NMR. Nine compounds were isolated and identified as
Chun-ying Xie et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(7), 1060-1062 (2011-11-10)
To study the chemical constituents of Pithecellobium clypearia. Silica gel column chromatography and Sephadex LH-20 were used to separate and purify compounds from the EtOAc soluble fraction of Pithecellobium clypearia. 9 compounds were elucidated on the basis of physicochemical properties
Qi-lei Guo et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 30(1), 35-37 (2007-06-02)
To study the chemical constituents of the root of Inula cappa. The compounds were isolated and purified by recrystallization and chromatography with silica gel column and Sephadex LH-20 column. Their structures were identified by physico-chemical properties and spectral analysis. Seven
K Halse et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 101(6), 430-436 (1993-06-01)
About 50 years ago crystalline deposits of a substance representing approximately 7 per cent wet tissue weight and believed to be hentriacontane or a mixture of similar long-chain n-alkanes (30-34 carbon atoms) were detected in two discoloured and swollen cow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service