Skip to Content
Merck
All Photos(1)

Key Documents

280828

Sigma-Aldrich

Terephthalic acid disodium salt

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CO2Na)2
CAS Number:
Molecular Weight:
210.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccc(cc1)C([O-])=O

InChI

1S/C8H6O4.2Na/c9-7(10)5-1-2-6(4-3-5)8(11)12;;/h1-4H,(H,9,10)(H,11,12);;/q;2*+1/p-2

InChI key

VIQSRHWJEKERKR-UHFFFAOYSA-L

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bashkim Misini et al.
Journal of neuroscience methods, 251, 1-6 (2015-05-17)
Terephthalate (TA(2-)), which reacts with highly reactive oxygen species (hROS) to form the fluorophor 2-hydroxy terephthalic acid (OH-TA) with a high selectivity, has been used for determining hROS formation during in vivo microdialysis. Previously this involved collecting fractions of the
Kazuhiko Takeda et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(4), 459-464 (2018-04-13)
A non-enzymatic fluorescence method for the determination of hydrogen peroxide (H2O2) was investigated. This method is based on the hydroxylation reaction of terephthalate (TP) by hydroxyl radical formed from reaction between H2O2 and Fe(II), resulting in the formation of a
Saliha Esin Çelik et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 33(3), 299-305 (2017-03-18)
In order to evaluate the antioxidant capacity/activity of β-carotene (BC) in aqueous media, we investigated the inclusion complexes of BC with methyl-β-cyclodextrin (Me-β-CD), 2-hydroxypropyl-β-cyclodextrin (HP-β-CD) and 2-hydroxyethyl-β-cyclodextrin (HE-β-CD) that enhance water solubility and chemical stability. The inclusion complexes (monitored by
Sébastien Moret et al.
Forensic science international, 291, 83-93 (2018-09-02)
Metal-Organic Frameworks (MOFs) are porous crystalline structures, currently used as sensors, separation membranes, and as catalysts. Due to their physicochemical and optical properties, they have been recently proposed for fingermark detection. This study further explored their potential for fingermark detection.
Aly Derbalah et al.
Chemosphere, 244, 125464-125464 (2019-12-04)
Reactive oxygen species (ROS) including singlet oxygen (1O2) and hydroxylradicals (OH) photogenerated in natural waters play important roles in indirect photolysis of man-made pollutants. This study was conducted to investigate how the generation of these two ROS influences the degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service