Skip to Content
Merck
All Photos(2)

Key Documents

277991

Sigma-Aldrich

Palladium on barium sulfate

reduced, extent of labeling: 5 wt. % loading

Synonym(s):

Pd/BaSO4, Rosenmund catalyst

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€46.10
10 G
€197.00
50 G
€487.00

€46.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€46.10
10 G
€197.00
50 G
€487.00

About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22

€46.10


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

5 wt. % loading

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for the hydrogenation, the Rosenmund reduction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bold, G.; Steiner, H. et al.
Helvetica Chimica Acta, 73, 405-405 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service