Skip to Content
Merck
All Photos(1)

Documents

277657

Sigma-Aldrich

Bis[(trimethylsilyl)tributyl]stannyl phosphate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]2P(O)OSn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
531.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

SMILES string

CCCC[Sn](CCCC)(CCCC)OP(=O)(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C6H19O4PSi2.3C4H9.Sn/c1-12(2,3)9-11(7,8)10-13(4,5)6;3*1-3-4-2;/h1-6H3,(H,7,8);3*1,3-4H2,2H3;/q;;;;+1/p-1

InChI key

ASQKAZSYRUOBEU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For the preparation of unesterified acyl or sugar phosphates under mild conditions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 17, 75-75 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service