Skip to Content
Merck
All Photos(1)

Key Documents

277312

Sigma-Aldrich

2-Propyl-1-pentanol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€57.70
10 G
€257.00

€57.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€57.70
10 G
€257.00

About This Item

Linear Formula:
(CH3CH2CH2)2CHCH2OH
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.43 (lit.)

bp

80-82 °C/12 mmHg (lit.)

density

0.83 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCC(CO)CCC

InChI

1S/C8H18O/c1-3-5-8(7-9)6-4-2/h8-9H,3-7H2,1-2H3

InChI key

LASHFHLFDRTERB-UHFFFAOYSA-N

Application

2-Propyl-1-pentanol may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service